Learn More
KCNN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KCNN2.
Brand: Abnova PAB27742.50ug
Description
Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. The protein encoded by this gene is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits. This gene is a member of the KCNN family of potassium channel genes. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| KCNN2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of KCNN2. | |
| In PBS (0.09% sodium azide) | |
| KCa2.2/SK2/SKCA2/hSK2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 3781 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| KCNN2 | |
| KCNN2 | |
| A synthetic peptide corresponding to 18 amino acid at C-terminus of human KCNN2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only