Learn More
KCNK17, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant KCNK17.
Brand: Abnova PAB24275.100uL
Description
The protein encoded by this gene belongs to the family of potassium channel proteins containing two pore-forming P domains. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. This gene is activated at alkaline pH. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: CSCCHHSSKEDFKSQSWRQGPDREPESHSPQQGCYPEGPMGIIQHLEPSAHAAGCGKDSSpecifications
| KCNK17 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant KCNK17. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| K2p17.1/TALK-2/TALK2/TASK-4/TASK4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 89822 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| KCNK17 | |
| KCNK17 | |
| Recombinant protein corresponding to amino acids of human KCNK17. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only