Learn More
KCNJ8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant KCNJ8.
Brand: Abnova PAB22503.100uL
Description
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins. [provided by RefSeq
Sequence: KRSPLYDISATDLANQDLEVIVILEGVVETTGITTQARTSYIAEEIQWGHRFVSIVTEEEGVYSVDYSKFGNTVKVAAPRCSARELDEKPSILIQTLQKSELSHQNSLRKRNSMRRNNSMRRNNSIRRNNSSLMVPKVQFMTPEGNQNTSpecifications
| KCNJ8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant KCNJ8. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIR6.1/uKATP-1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3764 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| KCNJ8 | |
| KCNJ8 | |
| Recombinant protein corresponding to amino acids of human KCNJ8. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only