Learn More
KCNJ12, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant KCNJ12.
Brand: Abnova PAB27489.100uL
Description
This gene encodes an inwardly rectifying K+ channel which may be blocked by divalent cations. This protein is thought to be one of multiple inwardly rectifying channels which contribute to the cardiac inward rectifier current (IK1). The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]
Sequence: RCSAKDLVENKFLLPSANSFCYENELAFLSRDEEDEADGDQDGRSRDGLSPQARHDFDRLQAGGGVLEQRPYRRESEISpecifications
| KCNJ12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant KCNJ12. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| FLJ14167/IRK2/KCNJN1/Kir2.2/Kir2.2v/hIRK/hIRK1/hkir2.2x/kcnj12x | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3768 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| KCNJ12 | |
| KCNJ12 | |
| Recombinant protein corresponding to amino acids of human KCNJ12. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only