Learn More
KCNG1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant KCNG1.
Brand: Abnova PAB23612.100uL
Description
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This gene is abundantly expressed in skeletal muscle. Multiple alternatively spliced transcript variants have been found in normal and cancerous tissues. [provided by RefSeq
Sequence: ELKQEQERVMFRRAQFLIKTKSQLSVSQDSDILFGSASSDTRDNNSpecifications
| KCNG1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant KCNG1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| K13/KCNG/KV6.1/MGC12878/kH2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3755 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| KCNG1 | |
| KCNG1 | |
| Recombinant protein corresponding to amino acids of human KCNG1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only