missing translation for 'onlineSavingsMsg'
Learn More

KATNB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant KATNB1.

Brand:  Abnova PAB23761.100uL

Product Code. 16196180

  • £535.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60kDa ATPase (p60 subunit A 1) and an 80kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. Katanin is a member of the AAA family of ATPases. [provided by RefSeq

Sequence: SPAMDVQFPVPNLEVLPRPPVVASTPAPKAEPAIIPATRNEPIGLKASDFLPAVKIPQQAELVDEDAMSQIRKGHDTMCVVLTSRHKNLDTVRAVWTMGDIKTSVDSAVAINDLSVV
Specifications

Specifications

KATNB1
Polyclonal
Rabbit polyclonal antibody raised against recombinant KATNB1.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
KAT
Rabbit
Antigen affinity purification
RUO
10300
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:1000-1:2500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
KATNB1
KATNB1
Recombinant protein corresponding to amino acids of human KATNB1.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only