missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa light chain, Rabbit, Clone: A21-Y, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of Human Kappa light chain.
Brand: Abnova MAB9775.100uL
This item is not returnable.
View return policy
Specifications
| Kappa light chain | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:500) The optimal working dilution should be determined by the end user. | |
| P01834 | |
| A synthetic peptide corresponding to N-terminus of human Human Kappa light chain. | |
| RUO | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| A21-Y | |
| Rabbit monoclonal antibody raised against synthetic peptide of Human Kappa light chain. | |
| In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide) | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Store at 4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction