missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANSL1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant KANSL1.
Brand: Abnova PAB20355.100uL
This item is not returnable.
View return policy
Description
Sequence: APLLERLSQLDSCVHPVLAFPDDVPTSLHFQSMLKSQWQNKPFDKIKPPKKLSLKHRAPMPGSLPDSARKDRHKLVSSFLTTAKLSHHQTRPDRTHRQHLDDVGAVPMVERVTAPKAERLLNPPPPVHDPNHSKMRLRDHSSSpecifications
| KANSL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant KANSL1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CENP-36/KDVS/KIAA1267/MSL1v1/NSL1/hMSL1v1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 284058 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| KANSL1 | |
| KANSL1 | |
| Recombinant protein corresponding to amino acids of human KANSL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UL KANSL1 polyclonal antibody
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate monobasic, potasssium phosphate dibasic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction