missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANK2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KANK2.
Brand: Abnova PAB27416.100ug
This item is not returnable.
View return policy
Specifications
| KANK2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| KANK2 | |
| KANK2 | |
| A synthetic peptide corresponding to 16 amino acids at C-terminus of human KANK2. | |
| 100 μg | |
| Primary | |
| Two alternatively spliced transcript variants encoding different isoforms have been identified. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of KANK2. | |
| In PBS (0.02% sodium azide) | |
| ANKRD25/DKFZp434N161/FLJ20004/KIAA1518/MGC119707/MXRA3/SIP | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 25959 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction