missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD2B (KDM4B) Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Kdm4b.
Brand: Abnova PAB15825.50ug
This item is not returnable.
View return policy
Specifications
| JMJD2B (KDM4B) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Kdm4b. | |
| In PBS (50% glycerol, 0.02% sodium azide) | |
| 4732474L06Rik/KDM4B/MGC6349/mKIAA0876 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Specific to recombinant protein GX2472. This antibody detects mJMJD2B protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Jmjd2b | |
| Jmjd2b | |
| Recombinant GST fusion protein corresponding to 156 amino acids of mouse Kdm4b. | |
| RUO | |
| 193796 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction