missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAK2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of JAK2.
Brand: Abnova PAB12770.100ug
This item is not returnable.
View return policy
Specifications
| JAK2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of JAK2. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| JAK2 | |
| A synthetic peptide corresponding to residues surrounding Y1007 and Y1008 of human, mouse and rat JAK2. | |
| RUO | |
| 3717/16452/24514 | |
| Human, Mouse, Rat | |
| Liquid |
| Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| JTK10 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes JAK2 without phosphorylated sites of Tyr1007/1008. It does not cross-react with phosphospecific peptide. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction