Learn More
JAK2 (phospho Y1007), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of JAK2.
Brand: Abnova PAB5741.100ug
Description
This gene product is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways. It has been found to be constituitively associated with the prolactin receptor and is required for responses to gamma interferon. Mice that do not express an active protein for this gene exhibit embryonic lethality associated with the absence of definitive erythropoiesis. [provided by RefSeq
Specifications
| JAK2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of JAK2. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| JTK10 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| JAK2 | |
| JAK2 | |
| Synthetic phosphopeptide corresponding to residues surrounding Y1007 of human JAK2. | |
| RUO | |
| 3717 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only