Learn More
ITGB2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ITGB2.
Brand: Abnova PAB12294.100ug
Description
The product of this gene belongs to the integrin beta chain family of proteins. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. This gene encodes the integrin beta chain beta 2. A given chain may combine with multiple partners resulting in different integrins. For example, beta 2 combines with the alpha L chain to form the integrin LFA-1, and combines with the alpha M chain to form the integrin Mac-1. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Defects in this gene are the cause of leukocyte adhesion deficiency type I (LAD1). Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq
Specifications
| ITGB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ITGB2. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| CD18/LAD/LCAMB/LFA-1/MAC-1/MF17/MFI7 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 3689 | |
| Human | |
| Lyophilized |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ITGB2 | |
| ITGB2 | |
| A synthetic peptide corresponding to N-terminus of human ITGB2. | |
| 100 μg | |
| Primary | |
| The synthetic peptide of this antibody different from the related mouse sequence by five amino acids. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only