missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGAM Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Itgam.
Brand: Abnova PAB12135.100uL
This item is not returnable.
View return policy
Specifications
| ITGAM | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Itgam. | |
| In PBS (30% glycerol, 0.09% sodium azide) | |
| CD11b/CD18/CR3/CR3A/Cd11b/F730045J24Rik/Ly-40/MAC1/Mac-1/Mac-1a | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunocytochemistry (1:200) Immunofluorescence (1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:400) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Itgam | |
| Itgam | |
| A synthetic peptide corresponding to amino acids 250-350 of mouse Itgam. | |
| RUO | |
| 16409 | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction