missing translation for 'onlineSavingsMsg'
Learn More

IRS2 (phospho S731), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of IRS2.

Brand:  Abnova PAB12629.100ug

Product Code. 16079996

  • £349.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes the insulin receptor substrate 2, a cytoplasmic signaling molecule that mediates effects of insulin, insulin-like growth factor 1, and other cytokines by acting as a molecular adaptor between diverse receptor tyrosine kinases and downstream effectors. The product of this gene is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation, as well as by an interleukin 4 receptor-associated kinase in response to IL4 treatment. [provided by RefSeq

Sequence: ASSPA
Specifications

Specifications

IRS2
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of IRS2.
In TBS, pH 7.2 (BSA, 10% Proclin300)
IRS2
Synthetic phosphopeptide corresponding to residues surrounding S731 of human IRS2.
RUO
8660
Human, Mouse, Rat
Liquid
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
IRS2
Rabbit
100 μg
Primary
This antibody recognizes ∼180 KDa of human IRS2 (pSer731).
Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only