missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRS1 (phospho S307), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of IRS1.
Brand: Abnova PAB10078.100ug
This item is not returnable.
View return policy
Specifications
| IRS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of IRS1. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| HIRS-1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Reactivity occurs against human IRS1 pS307 protein and This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated human IRS1 is minimal by ELISA. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:10000) Western Blot (1:250-1:1500) The optimal working dilution should be determined by the end user. | |
| IRS1 | |
| IRS1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S307 of human IRS1. | |
| RUO | |
| 3667 | |
| Bovine, Canine, Chicken, Human, Monkey, Mouse, Pig, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction