Learn More
IRF3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IRF3.
Brand: Abnova PAB24842.100uL
Description
IRF3 encodes interferon regulatory factor 3, a member of the interferon regulatory transcription factor (IRF) family. IRF3 is found in an inactive cytoplasmic form that upon serine/threonine phosphorylation forms a complex with CREBBP. This complex translocates to the nucleus and activates the transcription of interferons alpha and beta, as well as other interferon-induced genes. [provided by RefSeq
Specifications
| IRF3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| IRF3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3661 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of IRF3. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| IRF3 | |
| A synthetic peptide corresponding to IRF3. | |
| 100 μL | |
| Primary | |
| IRF3 polyclonal antibody detects endogenous levels of IRF3 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only