Learn More
IRF2BP2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IRF2BP2.
Brand: Abnova PAB25709.100ug
Description
This gene encodes an interferon regulatory factor-2 (IRF2) binding protein that interacts with the C-terminal transcriptional repression domain of IRF2. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Specifications
| IRF2BP2 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| IRF2BP2 | |
| IRF2BP2 | |
| A synthetic peptide corresponding to 17 amino acids at N-terminus of human IRF2BP2. | |
| 100 μg | |
| Primary | |
| At least two isoforms of IRF2BP2 are known to exist; this antibody will detect both. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of IRF2BP2. | |
| In PBS (0.05% sodium azide) | |
| MGC72189 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 359948 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only