Learn More
INTS6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant INTS6
Brand: Abnova PAB28493.100uL
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. The protein encoded by this gene is a DEAD box protein that is part of a complex that interacts with the C-terminus of RNA polymerase II and is involved in 3' end processing of snRNAs. In addition, this gene is a candidate tumor suppressor and located in the critical region of loss of heterozygosity (LOH). Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]
Sequence: VVNNHIGGKGPPAPTTQAQPDLIKPLPLHKISETTNDSIIHDVVENHVADQLSSDITPNAMDTEFSASSPASLLERPTNHMEALGHDHLGTNDLTVGGFLENHEEPRDKEQCAEENIPASSLNKGKKLMHCRSHEEVNTELKAQSpecifications
| INTS6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant INTS6 | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| DBI-1/DDX26/DDX26A/DICE1/DKFZp434B105/HDB/INT6/Notchl2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 26512 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:50-1:200) Western Blot(1:250 - 1:500) Immunofluorescence(1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| INTS6 | |
| INTS6 | |
| Recombinant protein corresponding to amino acids of human INTS6 | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only