Learn More
IMPDH2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IMPDH2.
Brand: Abnova PAB28430.100uL
Beschreibung
This gene encodes the rate-limiting enzyme in the de novo guanine nucleotide biosynthesis. It is thus involved in maintaining cellular guanine deoxy- and ribonucleotide pools needed for DNA and RNA synthesis. The encoded protein catalyzes the NAD-dependent oxidation of inosine-5'-monophosphate into xanthine-5'-monophosphate, which is then converted into guanosine-5'-monophosphate. This gene is up-regulated in some neoplasms, suggesting it may play a role in malignant transformation. [provided by RefSeq]
Sequence: GFITDPVVLSPKDRVRDVFEAKARHGFCGIPITDTGRMGSRLVGIISSRDIDFLKEEEHDCFLEEIMTKREDLVVAPAGITLKEANEILQRSKKGKLPIVNEDDELVAIIARTDLKKNRDYPLASKDSpezifikation
| IMPDH2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant IMPDH2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| IMPD2/IMPDH-II | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3615 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:200-1:500) Western Blot(1:100-1:250) The optimal working dilution should be determined by the end user. | |
| IMPDH2 | |
| IMPDH2 | |
| Recombinant protein corresponding to amino acids of recombinant IMPDH2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only