Learn More
ILK (phospho T173), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ILK.
Brand: Abnova PAB1640.400uL
Description
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| ILK | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of ILK. | |
| In PBS (0.09% sodium azide) | |
| DKFZp686F1765/P59 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3611 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA | |
| Unconjugated | |
| Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
| ILK | |
| ILK | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T173 of human ILK. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only