Learn More
ILF3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ILF3.
Brand: Abnova PAB28534.100uL
Description
This gene encodes a double-stranded RNA (dsRNA) binding protein that complexes with other proteins, dsRNAs, small noncoding RNAs, and mRNAs to regulate gene expression and stabilize mRNAs. This protein was first discovered to be a subunit of the nuclear factor of activated T-cells (NFAT); a transcription factor required for T-cell expression of interleukin 2. NFAT is a heterodimer of 45kDa and 90kDa proteins, the larger of which is the product of this gene. These proteins have been shown to affect the redistribution of nuclear mRNA to the cytoplasm. Knockdown of NF45 or NF90 protein retards cell growth; possibly by inhibition of mRNA stabilization. In contrast, an isoform (NF110) of this gene that is predominantly restricted to the nucleus has only minor effects on cell growth when its levels are reduced. Alternative splicing results in multiple transcript variants encoding distinct isoforms
Sequence: VMAKHSSVYPTQEELEAVQNMVSHTERALKAVSDWIDEQEKGSSEQAESDNMDVPPEDDSKEGAGEQKTEHMTRTLRGVMRVGLVAKGLLLKGDLDLELVLLCKEKPTTALLDKVADNLAIQLAAVTEDKYEILQSVDDAAIVIKSpecifications
| ILF3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ILF3. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| CBTF/DRBF/DRBP76/MMP4/MPHOSPH4/MPP4/NF-AT-90/NF110/NF90/NFAR/NFAR-1/NFAR2/TCP110/TCP80 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3609 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:500) Immunofluorescence (1-4 ug/ml) Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| ILF3 | |
| ILF3 | |
| Recombinant protein corresponding to amino acids of human ILF3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only