missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Il8rb, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Il8rb.
Marca: Abnova PAB8942.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| Il8rb | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Il8rb. | |
| In buffer containing 0.02% sodium azide | |
| Il8rb | |
| A synthetic peptide corresponding to C-terminus of rat/mouse Il8rb. | |
| RUO | |
| 12765/29385 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:500) Immunoprecipitation (1:250) The optimal working dilution should be determined by the end user. | |
| CD128/CDw128/CXCR2/Cmkar2/Gpcr16/IL-8Rh/IL-8rb/IL8RA/mIL-8RH | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction