Learn More
IL34 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL34.
Brand: Abnova PAB13397.100ug
Description
Interleukin-34 is a cytokine that promotes the differentiation and viability of monocytes and macrophages through the colony-stimulating factor-1 receptor (CSF1R; MIM 164770) (Lin et al., 2008 [PubMed 18467591]).[supplied by OMIM
Specifications
| IL34 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of IL34. | |
| In PBS (0.02% sodium azide) | |
| C16orf77/IL-34/MGC34647 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| IL34 | |
| IL34 | |
| A synthetic peptide corresponding to N-terminus 16 amino acids of human IL34. | |
| RUO | |
| 146433 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only