Learn More
IL32 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL32.
Brand: Abnova PAB13186.100ug
Description
This gene encodes a member of the cytokine family. The protein contains a tyrosine sulfation site, 3 potential N-myristoylation sites, multiple putative phosphorylation sites, and an RGD cell-attachment sequence. Expression of this protein is increased after the activation of T-cells by mitogens or the activation of NK cells by IL-2. This protein induces the production of TNFalpha from macrophage cells. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
| IL32 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of IL32. | |
| In PBS (0.02% sodium azide) | |
| IL-32alpha/IL-32beta/IL-32delta/IL-32gamma/NK4/TAIF/TAIFa/TAIFb/TAIFc/TAIFd | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects the largest isoform of IL-32. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (2.5-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| IL32 | |
| IL32 | |
| A synthetic peptide corresponding to internal region 16 amino acids of human IL32. | |
| RUO | |
| 9235 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only