Learn More
IL3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against IL3.
Brand: Abnova PAB16166.100ug
Description
The protein encoded by this gene is a potent growth promoting cytokine. This cytokine is capable of supporting the proliferation of a broad range of hematopoietic cell types. It is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. This cytokine has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders. [provided by RefSeq
Specifications
| IL3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against IL3. | |
| Lyophilized from PBS. | |
| IL-3/MCGF/MGC79398/MGC79399/MULTI-CSF | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1-2 ug/mL) Western Blot (2-10 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| IL3 | |
| IL3 | |
| Human IL3. | |
| RUO | |
| 3562 | |
| Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only