Learn More
IL18 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL18.
Brand: Abnova PAB18297.100ug
Description
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein (IL18BP) can specifically interact with this cytokine, and thus negatively regulate its biological activity. [provided by RefSeq
Specifications
| IL18 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| IL18 | |
| IL18 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human IL18. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
| Western Blot | |
| 0.5 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of IL18. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| IGIF/IL-18/IL-1g/IL1F4/MGC12320 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 3606 | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only