Learn More
IL17RD, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IL17RD.
Brand: Abnova PAB16178.100ug
Description
Fibroblast growth factors (FGFs; see MIM 603726) are secreted proteins involved in cellular proliferation, migration, differentiation, and survival. FGF activity is negatively regulated by members of the 'sprouty' family (e.g., SPRY1, MIM 602465). The SEF protein is a modulator of FGF signaling.[supplied by OMIM
Specifications
| IL17RD | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant IL17RD. | |
| Lyophilized from PBS. | |
| DKFZp434N1928/FLJ35755/IL-17RD/IL17RLM/MGC133309/SEF | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
| ELISA | |
| Unconjugated | |
| ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| IL17RD | |
| IL17RD | |
| Recombinant protein corresponding to human IL17RD. | |
| RUO | |
| 54756 | |
| Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only