Learn More
IL10RA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL10RA.
Brand: Abnova PAB18395.100ug
Description
The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene. [provided by RefSeq
Specifications
| IL10RA | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| IL10RA | |
| IL10RA | |
| A synthetic peptide corresponding to residues surrounding Y496 of human IL10RA. | |
| 100 μg | |
| Primary | |
| This antibody is specific to IL10RA. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of IL10RA. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| CDW210A/HIL-10R/IL-10R1/IL10R | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 3587 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only