missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKBKA/IKBKB, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IKBKA/IKBKB.
Brand: Abnova PAB25508.100ug
This item is not returnable.
View return policy
Description
Sequence: CTSFVSpecifications
| IKBKA/IKBKB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of IKBKA/IKBKB. | |
| In PBS, pH 7.4 (1% BSA, 50% glycerol, 0.02% sodium azide) | |
| IKBKA/IKK-alpha/IKK1/IKKA/NFKBIKA/TCF16 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 1147/3551 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| O15111;O14920 | |
| CHUK | |
| A synthetic peptide corresponding to residues surrounding S180/S181 human IKBKA/IKBKB. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction