missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yersinia pseudotuberculosis, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Yersinia pseudotuberculosis.
Brand: Abnova PAB13574.200uL
This item is not returnable.
View return policy
Specifications
| Yersinia pseudotuberculosis | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Yersinia pseudotuberculosis. | |
| Lyophilized from serum | |
| Yersinia pseudotuberculosis | |
| RUO | |
| Marmoset | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Immunocytochemistry (1:500) Immunoflourescence (1:500) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction