Learn More
HVCN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HVCN1.
Brand: Abnova PAB23414.100uL
Description
HVCN1 is a voltage-gated proton channel highly expressed in immune tissues. Channels like HVCN1 mediate the proton conductances required by phagocytic leukocytes for the oxidative burst that underlies microbial killing (Ramsey et al., 2006 [PubMed 16554753]).[supplied by OMIM
Sequence: GIIISVKTRSERQLLRLKQMNVQLAAKIQHLEFSCSEKEQEIERLNKLLRQHGLLGEVNSpecifications
| HVCN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HVCN1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| HV1/MGC15619/VSOP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 84329 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| HVCN1 | |
| HVCN1 | |
| Recombinant protein corresponding to amino acids of human HVCN1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only