Learn More
Rabbit anti-Human IgM Fc Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69700
245.65 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18611609
  • £350.00 / 10mg
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Human samples. It is validated for Western Blot, ELISA, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgM Fc
Polyclonal
Unconjugated
Rabbit
Ion exchange chromatography
RUO
Secondary
Human
Lyophilized
Western Blot, ELISA, Dot Blot
LYOPH
Lyophilized from PBS (pH 7.2)
Purified homogenous IgM isolated from human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
10 mg
B Cell Development and Differentiation Markers, Immunology
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.