Learn More
Rabbit anti-Human IgG1 Fc Secondary Antibody (245), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-40578H
Product Code. 30239677
  • £377.00 / 0.10mL
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG1 Fc
Monoclonal
HRP
Rabbit
Protein A purified
RUO
Human
Purified
ELISA
245
PBS
This antibody was obtained from a rabbit immunized with purified, recombinant Human IgG1 Fc (Uniprot#: P01857-1; Glu99-Lys330,C103S). HEK293 used as expression system.
0.1 mL
Secondary
Store at 4°C in the dark.