Learn More
Invitrogen™ Rabbit anti-Human Ig (Light chain) Recombinant Secondary Antibody
Rabbit Recombinant Monoclonal Secondary Antibody
Brand: Invitrogen™ MA527942
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human Ig (Light chain) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| P0CG04 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3537 | |
| -20°C | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| RM129 | |
| Unconjugated | |
| IGLC1 | |
| IGLC1 | |
| Human IgG. | |
| 100 μg | |
| Secondary | |
| Human | |
| Secondary Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.