missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG (H+L) Secondary Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-75070PCP
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
Specifications
| IgG (H+L) | |
| Polyclonal | |
| PBS | |
| This Rabbit anti-Human IgG (H+L) Secondary Antibody was developed against purified human IgG (H&L). | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay (CIA) | |
| PerCP | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction