Learn More
Rabbit anti-Human IgG Fc gamma Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen SA1600
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. SA1-600 reacts with human IgG and not with human IgA, IgM, kappa or lambda chains. SA1-600 has been successfully used in ELISA and Immunoelectrophoresis applications. The SA1-600 immunogen is purified human IgG, Fc gamma fragment. Country of Origin: USA
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Human IgG Fc gamma | |
| Polyclonal | |
| Unconjugated | |
| Purified human IgG, Fc gamma fragment. | |
| 2 mg | |
| Secondary | |
| 4° C | |
| IgG |
| ELISA, Immunoelectrophoresis | |
| 2.3 mg/mL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Human | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.