missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgE (Heavy chain) Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen PA516396
This item is not returnable.
View return policy
Description
PA5-16396 targets Human IgE (epsilon-Heavy Chain) in IHC (P) and FACS applications and shows reactivity with Human samples. The PA5-16396 immunogen is igE isolated from a pool of normal human sera.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgE (Heavy chain) | |
| Polyclonal | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| IgE isolated from a pool of normal human sera | |
| 500 μL | |
| Secondary | |
| 4° C | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Ig fraction | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction