missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit anti-Human IgD (Heavy chain) Secondary Antibody
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen™ PA516400
This item is not returnable.
View return policy
Description
PA5-16400 targets Human IgD (delta-Heavy Chain) in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-16400 immunogen is igD isolated from a pool of normal human sera.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgD (Heavy chain) | |
| Polyclonal | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| IgD isolated from a pool of normal human sera. | |
| 500 μL | |
| Secondary | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Ig fraction | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction