Learn More
HSPE1 Rabbit anti-Bovine, Dog, Frog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HSPE1.
Brand: Abnova PAB15431.100ug
Description
This gene encodes a major heat shock protein which functions as a chaperonin. Its structure consists of a heptameric ring which binds to another heat shock protein in order to form a symmetric, functional heterodimer which enhances protein folding in an ATP-dependent manner. This gene and its co-chaperonin, HSPD1, are arranged in a head-to-head orientation on chromosome 2. [provided by RefSeq
Specifications
| HSPE1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HSPE1. | |
| In PBS, pH 7.3 (0.02% sodium azide) | |
| CPN10/GROES/HSP10 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects ∼ 10KDa. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| HSPE1 | |
| HSPE1 | |
| A synthetic peptide corresponding to amino acids 91-101 of human HSPE1. | |
| RUO | |
| 3336 | |
| Bovine, Canine, Frog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only