Learn More
HSPA12B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HSPA12B.
Brand: Abnova PAB20831.100uL
Description
The protein encoded by this gene contains an atypical heat shock protein 70 (Hsp70) ATPase domain and is therefore a distant member of the mammalian Hsp70 family. This gene may be involved in susceptibility to atherosclerosis. [provided by RefSeq
Sequence: LAVPEMGLQGLYIGSSPERSPVPSPPGSPRTQESCGIAPLTPSQSPKPEVRAPQQASFSVVVAIDFGTTSSGYAFSFASDPEAIHMMRKWEGGSpecifications
| HSPA12B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HSPA12B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| C20orf60/FLJ32150/MGC131912/dJ1009E24.2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 116835 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| HSPA12B | |
| HSPA12B | |
| Recombinant protein corresponding to amino acids of human HSPA12B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only