Learn More
HSD17B8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HSD17B8.
Brand: Abnova PAB8493.25ug
Description
In mice, the Ke6 protein is a 17-beta-hydroxysteroid dehydrogenase that can regulate the concentration of biologically active estrogens and androgens. It is preferentially an oxidative enzyme and inactivates estradiol, testosterone, and dihydrotestosterone. However, the enzyme has some reductive activity and can synthesize estradiol from estrone. The protein encoded by this gene is similar to Ke6 and is a member of the short-chain dehydrogenase superfamily. An alternatively spliced transcript of this gene has been detected, but the full-length nature of this variant has not been determined. [provided by RefSeq
Specifications
| HSD17B8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HSD17B8. | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| D6S2245E/FABG/FABGL/H2-KE6/HKE6/KE6/RING2/SDR30C1/dJ1033B10.9 | |
| Rabbit | |
| 25 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| HSD17B8 | |
| HSD17B8 | |
| A synthetic peptide corresponding to human HSD17B8. | |
| RUO | |
| 7923 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only