Learn More
HSD17B4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HSD17B4.
Brand: Abnova PAB21385.100uL
Description
The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. [provided by RefSeq
Sequence: ESCEENGGLFEVGAGWIGKLRWERTLGAIVRQKNHPMTPEAVKANWKKICDFENASKPQSIQESTGSIIEVLSKSpecifications
| HSD17B4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HSD17B4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DBP/MFE-2/SDR8C1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3295 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| HSD17B4 | |
| HSD17B4 | |
| Recombinant protein corresponding to amino acids of human HSD17B4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only