Learn More
HS6ST2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HS6ST2.
Brand: Abnova PAB22628.100uL
Description
Heparan sulfate proteoglycans are ubiquitous components of the cell surface, extracellular matrix, and basement membranes, and interact with various ligands to influence cell growth, differentiation, adhesion, and migration. This gene encodes a member of the heparan sulfate (HS) sulfotransferase gene family, which catalyze the transfer of sulfate to HS. Different family members and isoforms are thought to synthesize heparan sulfates with tissue-specific structures and functions. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: RKRQEQRKFLKGRLLQTHFQSQGQGQSQNPNQNQSQNPNPNANQNLTQNLMQNLTQSLSQKENRESPKQNSGKEQNDNTSNGTNDSpecifications
| HS6ST2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HS6ST2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC130022/MGC130023 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 90161 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| HS6ST2 | |
| HS6ST2 | |
| Recombinant protein corresponding to amino acids of human HS6ST2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only