Learn More
HOXC11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HOXC11.
Brand: Abnova PAB23260.100uL
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene binds to a promoter element of the lactase-phlorizin hydrolase. It also may play a role in early intestinal development. An alternatively spliced variant encoding a shorter isoform has been described but its full-length nature has not been determined. [provided by RefSeq
Sequence: STVSSFLPQAPSRQISYPYSAQVPPVREVSYGLEPSGKWHHRNSYSSCYAAADELMHRECLPPSTVTEILMKNEGSYGGHHHPSAPHATPAGFYSSVNKNSVLPSpecifications
| HOXC11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HOXC11. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| HOX3H/MGC4906 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3227 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| HOXC11 | |
| HOXC11 | |
| Recombinant protein corresponding to amino acids of human HOXC11. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only