Learn More
HOXA1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HOXA1.
Brand: Abnova PAB19786.100ug
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region. [provided by RefSeq
Specifications
| HOXA1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HOXA1. | |
| In serum (0.5% BSA, 0.02% sodium azide) | |
| BSAS/HOX1/HOX1F/MGC45232 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 3198 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:20000-1:80000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user. | |
| HOXA1 | |
| HOXA1 | |
| A synthetic peptide corresponding to 15 amino acids at internal region of human HOXA1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only