Learn More
HNRPLL Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HNRPLL.
Brand: Abnova PAB24468.100uL
Description
HNRNPLL is a master regulator of activation-induced alternative splicing in T cells. In particular, it alters splicing of CD45 (PTPRC; MIM 151460), a tyrosine phosphatase essential for T-cell development and activation (Oberdoerffer et al., 2008 [PubMed 18669861]).[supplied by OMIM
Sequence: QASKNIIQPPSCVLHYYNVPLCVTEETFTKLCNDHEVLTFIKYKVFDAKPSAKTLSGLLEWECKTDSpecifications
| HNRPLL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HNRPLL. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| SRRF | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 92906 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| HNRPLL | |
| HNRPLL | |
| Recombinant protein corresponding to amino acids of human HNRPLL. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only