Learn More
HNRNPR, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HNRNPR.
Brand: Abnova PAB21792.100uL
Description
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs and also contains a nuclear localization motif. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: MANQVNGNAVQLKEEEEPMDTSSVTHTEHYKTLIEAGLPQKVAERLDEIFQTSpecifications
| HNRNPR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HNRNPR. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ25714/HNRPR/hnRNP-R | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10236 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| HNRNPR | |
| HNRNPR | |
| Recombinant protein corresponding to amino acids of human HNRNPR. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only