missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hmgb3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Hmgb3.
Brand: Abnova PAB8488.25ug
This item is not returnable.
View return policy
Specifications
| Hmgb3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Hmgb3. | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| Hmg2a/Hmg4 | |
| Rabbit | |
| 25 μg | |
| Primary | |
| Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| Hmgb3 | |
| Hmgb3 | |
| A synthetic peptide corresponding to mouse Hmgb3. | |
| RUO | |
| 15354 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction