missing translation for 'onlineSavingsMsg'
Learn More
Learn More
histone H2B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of histone H2B.
Brand: Abnova PAB16672.50uL
This item is not returnable.
View return policy
Description
Sequence: SAAEKKPASKAPAGKASpecifications
| histone H2B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of histone H2B. | |
| In antiserum (0.05% sodium azide) | |
| htb1 | |
| A synthetic peptide corresponding to Schizosaccharomyces pombe histone H2B. | |
| RUO | |
| 2539523 | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| ChIP Assay, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| htb1 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| S. pombe | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction